CAS RN 198821-22-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Merimepodib, MedChemExpress
MedChemExpress Merimepodib (VX-497) is a noncompetitive and oral inhibitor of inosine monophosphate dehydrogenase (IMPDH) with broad spectrum antiviral activities.