CAS RN 1984862-48-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
OABK hydrochloride, MedChemExpress
MedChemExpress OABK hydrochloride is a small-molecule switch that can be used to control protein activity.