CAS RN 198474-05-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PF-06840003, MedChemExpress
MedChemExpress PF-06840003 (EOS200271) is a highly selective orally bioavailable IDO-1 inhibitor with IC50s of 0.41 μM, 0.59 μM, and 1.5 μM for hIDO-1, dIDO-1, and mIDO-1, respectively.