CAS RN 1955550-51-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ FMK 9a
Potent ATG4B/autophagin-1 inhibitor; also inhibits calpain and cathepsin B
2
FMK 9a, MedChemExpress
MedChemExpress FMK 9a is an autophagin-1 inhibitor with IC50 values of 80 and 73 μM in FRET and LRA assay.