CAS RN 1942919-79-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NVP-BSK805 dihydrochloride, MedChemExpress
MedChemExpress NVP-BSK805 dihydrochloride is an ATP-competitive JAK2 inhibitor, with IC50s of 0.48 nM, 31.63 nM, 18.68 nM, and 10.76 nM for JAK2 JH1 (JAK homology 1), JAK1 JH1, JAK3 JH1, and TYK2 JH1, respectively.