CAS RN 194093-42-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pumosetrag Hydrochloride, MedChemExpress
MedChemExpress Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.