CAS RN 1936529-65-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YKL-05-099, MedChemExpress
MedChemExpress YKL-05-099 is a salt-inducible kinase (SIK) inhibitor. YKL-05-099 binds to SIK1 and SIK3 with IC50s of ∼10 and ∼30 nM, respectively. YKL-05-099 has slightly less potent SIK2-inhibitory (IC50=40 nM).