CAS RN 193000-39-4
IUPAC Nombre:
5-{[7-(benzyloxy)quinazolin-4-yl]amino}-4-fluoro-2-methylphenol hydrochloride
Sinónimos:
5-{[7-(benzyloxy)quinazolin-4-yl]amino}-4-fluoro-2-methylphenol hydrochloride
Peso molecular (g/mol):
411.86
Fórmula molecular:
C22H19ClFN3O2
InChi Key:
AVRHWGLIYGJSOD-UHFFFAOYSA-N
SMILES:
Cl.CC1=CC(F)=C(NC2=C3C=CC(OCC4=CC=CC=C4)=CC3=NC=N2)C=C1O
1
–
2
de
2
resultados
1
Tocris Bioscience™ ZM 323881 hydrochloride
CAS: 193000-39-4 Fórmula molecular: C22H19ClFN3O2
2
ZM323881 hydrochloride, MedChemExpress
MedChemExpress ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM.