CAS RN 192185-71-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tipifarnib (S enantiomer), MedChemExpress
MedChemExpress Tipifarnib S enantiomer is the S-enantiomer of Tipifarnib. Tipifarnib is a potent and specific farnesyltransferase (FTase) inhibitor with IC50 of 0.6 nM. Tipifarnib S enantiomer is the less active isomer.