CAS RN 189950-66-1
IUPAC Nombre:
4-{[1-({2-carboxy-1-[(4-nitrophenyl)carbamoyl]ethyl}carbamoyl)-2-methylpropyl]carbamoyl}-4-(3-carboxy-2-acetamidopropanamido)butanoic acid
Sinónimos:
4-{[1-({2-carboxy-1-[(4-nitrophenyl)carbamoyl]ethyl}carbamoyl)-2-methylpropyl]carbamoyl}-4-(3-carboxy-2-acetamidopropanamido)butanoic acid
Peso molecular (g/mol):
638.59
Fórmula molecular:
C26H34N6O13
InChi Key:
GGXRLUDNGFFUKI-UHFFFAOYNA-N
SMILES:
CC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(C)=O)C(=O)NC(CC(O)=O)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
1
–
5
de
5
resultados
2
MedChemExpress Ac-DEVD-pNA
MedChemExpress Ac-DEVD-pNA is a colorimetric substrate for caspase-3 (CPP32) and related cysteine proteases.
3
MedChemExpress Ac-DEVD-pNA
MedChemExpress Ac-DEVD-pNA is a colorimetric substrate for caspase-3 (CPP32) and related cysteine proteases.
4
MedChemExpress Ac-DEVD-pNA
MedChemExpress Ac-DEVD-pNA is a colorimetric substrate for caspase-3 (CPP32) and related cysteine proteases.
5
MedChemExpress Ac-DEVD-pNA
MedChemExpress Ac-DEVD-pNA is a colorimetric substrate for caspase-3 (CPP32) and related cysteine proteases.