CAS RN 189699-82-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FR-188582, MedChemExpress
MedChemExpress FR-188582 is a highly selective inhibitor of cyclooxygenase (COX)-2, with an IC50 value of 17 nM.