CAS RN 189388-22-5
IUPAC Nombre:
2-({1-[2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl}formamido)-N-(1-carbamoyl-2-phenylethyl)-3-(1H-indol-3-yl)propanamide
Sinónimos:
endomorphin-1
Peso molecular (g/mol):
610.72
Fórmula molecular:
C34H38N6O5
InChi Key:
ZEXLJFNSKAHNFH-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=C(O)C=C1)C(=O)N1CCCC1C(=O)NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CC1=CC=CC=C1)C(N)=O
1
–
4
de
4
resultados
1
MedChemExpress Endomorphin 1
MedChemExpress Endomorphin 1, a high affinity, highly selective agonist of the μ-opioid receptor, displays reasonable affinities for kappa3 binding sites, with Ki value between 20 and 30 nM.
2
MedChemExpress Endomorphin 1
MedChemExpress Endomorphin 1, a high affinity, highly selective agonist of the μ-opioid receptor, displays reasonable affinities for kappa3 binding sites, with Ki value between 20 and 30 nM.
3
MedChemExpress Endomorphin 1
MedChemExpress Endomorphin 1, a high affinity, highly selective agonist of the μ-opioid receptor, displays reasonable affinities for kappa3 binding sites, with Ki value between 20 and 30 nM.