CAS RN 189362-06-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
K134, MedChemExpress
MedChemExpress K134 is a phosphodiesterase 3 (PDE3) inhibitor. The IC50s of K134 toward PDE3A, PDE3B, PDE5, PDE2 and PDE4 are 0.1, 0.28, 12.1, >300 and >300 μM, respectively.