CAS RN 1889279-16-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ A 485
Histone Acetyltransferase Inhibitors
2
A-485, MedChemExpress
MedChemExpress A-485 is a potent and selective catalytic inhibitor of p300/CBP with IC50s of 9.8 nM and 2.6 nM for p300 and CBP histone acetyltransferase (HAT), respectively.
3
4