CAS RN 1883602-20-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TES-991, MedChemExpress
MedChemExpress TES-991 is a potent and selective human α‐Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM.