CAS RN 187870-78-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Rimeporide, MedChemExpress
MedChemExpress Rimeporide (EMD-87580) is a potent and selective inhibitor of the Na+/H+ exchanger (NHE-1).