CAS RN 1876467-74-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
BAY-1895344, TRC
CAS: 1876467-74-1 Fórmula molecular: C20H21N7O Sinónimo: 2-[(3R)-3-Metil-4-morfolinil]-4-(1-metil-1H-pirazol-5-il)-8-(1H-pirazol-3-il)-1,7-naftiridina∣BAY-1895344
2
Elimusertib, MedChemExpress
MedChemExpress Elimusertib (BAY-1895344) is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib has anti-tumor activity. Elimusertib can be used for the research of solid tumors and lymphomas.