CAS RN 18711-16-5
IUPAC Nombre:
6,7-dichloro-3-(hydroxyamino)-2H-indol-2-one
Sinónimos:
6,7-dichloro-3-(hydroxyamino)indol-2-one
Peso molecular (g/mol):
231.03
Fórmula molecular:
C8H4Cl2N2O2
InChi Key:
CVOUSAVHMDXCKG-UHFFFAOYSA-N
SMILES:
ONC1=C2C=CC(Cl)=C(Cl)C2=NC1=O
1
–
2
de
2
resultados
1
Tocris Bioscience™ NS 309
CAS: 18711-16-5 Fórmula molecular: C8H4Cl2N2O2
2
NS309, MedChemExpress
MedChemExpress NS309 is a potent and selective activator of the Ca2+-activated SK/IK potassium channels, but displays no activity at BK channels.