CAS RN 1851373-36-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
QC6352, MedChemExpress
MedChemExpress QC6352 is an orally available, selective and potent KDM4C inhibitor with an IC50 of 35 nM.