CAS RN 1848233-58-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DO34, MedChemExpress
MedChemExpress DO34 is a highly potent, selective and centrally active diacylglycerol lipase (DAGL) inhibitor, with an IC50 of 6 nM for DAGLα conversion of SAG to 2-AG, and an IC50 for DAGLβ.