CAS RN 181885-68-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress LDS-751
MedChemExpress LDS-751 is a nucleic acid stain principally detecting DNA. LDS-751 has high affinity for DNA and undergoes fluorescence enhancement upon binding, but with maximal emission at 670 nm.