Learn More
MedChemExpress LDS-751
Descripción
LDS-751 and Thiazole orange are used to differentiate erythrocytes, platelets, reticulocytes, and nucleated cells, and both can be excited at 488 nm.
Especificaciones
Especificaciones
| Contenido y almacenamiento | -20°C, sealed storage, away from moisture and light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Cantidad | 5 mg |
| Condiciones de envío | Room temperature |
| Solubilidad | DMSO : 83.33 mg/mL (176.55 mM; Need ultrasonic) |
| Objetivo | HBV |
| Tipo de producto | Dye Reagents |
| Peso molecular | 471.98 g/mol |
| CAS | 181885-68-7 |