CAS RN 1803274-65-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BAY-1436032, MedChemExpress
MedChemExpress BAY-1436032 is a novel pan-mutant isocitrate dehydrogenase 1 (IDH1) inhibitor.