CAS RN 1801873-49-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
OICR-0547, MedChemExpress
MedChemExpress OICR-0547 is a closely related derivative of OICR-9429. OICR-9429 is a novel small-molecule antagonist of the Wdr5-MLL interaction, while OICR-0547 is an inactive control compound that no longer binds to WDR5.