CAS RN 1801747-42-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SHP099, MedChemExpress
MedChemExpress SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC50 of 70 nM.