CAS RN 1796565-52-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Orexin 2 Receptor Agonist, MedChemExpress
MedChemExpress Orexin 2 Receptor Agonist is a potent (EC50 on OX2R is 23 nM) and OX2R-selective (OX1R/OX2R EC50 ratio is 70) agonist.