CAS RN 1794789-93-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Vilazodone-d8, MedChemExpress
MedChemExpress Vilazodone D8 is the a deuterium labeled vilazodone, which is a combined serotonin specific reuptake inhibitor (SSRI) and 5-HT1A receptor partial agonist.