CAS RN 179411-94-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SDZ 220-581 Ammonium salt, MedChemExpress
MedChemExpress SDZ 220-581 Ammonium salt is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.