CAS RN 178616-26-7
IUPAC Nombre:
1-(4-aminophenyl)-7,8-dimethoxy-4,5-dihydro-3H-2,3-benzodiazepin-4-one
Sinónimos:
1-(4-aminophenyl)-7,8-dimethoxy-3,5-dihydro-2,3-benzodiazepin-4-one
Peso molecular (g/mol):
311.34
Fórmula molecular:
C17H17N3O3
InChi Key:
MJKADKZSYQWGLL-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C=C2C(CC(=O)NN=C2C2=CC=C(N)C=C2)=C1
1
–
1
de
1
resultados
1
CFM-2, MedChemExpress
MedChemExpress CFM-2 is a potent and selective non-competitive AMPAR antagonist. CFM-2 possesses anticonvulsant activity in various models of seizures.