CAS RN 1784751-18-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TA-01, MedChemExpress
MedChemExpress TA-01 is a potent CK1 and p38 MAPK inhibitor, with IC50s of 6.4 nM, 6.8 nM, 6.7 nM for CK1ε, CK1δ and p38 MAPK, respectively. TA-01 acts as a cardiogenic inhibitor.