CAS RN 1783816-74-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ NVS PAK1 1
Potent and selective PAK1 inhibitor
2
NVS-PAK1-1, MedChemExpress
MedChemExpress NVS-PAK1-1 is a potent and selective allosteric PAK1 inhibitor with an IC50 of 5 nM.