CAS RN 176591-49-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Urocortin, human
MedChemExpress Urocortin, human, a 40-aa neuropeptide, acts as a selective agonist of endogenous CRF2 receptor, with Kis of 0.4, 0.3, and 0.5 nM for hCRF1, rCRF2α and mCRF2β, respectively.
2
MedChemExpress Urocortin, human
MedChemExpress Urocortin, human, a 40-aa neuropeptide, acts as a selective agonist of endogenous CRF2 receptor, with Kis of 0.4, 0.3, and 0.5 nM for hCRF1, rCRF2α and mCRF2β, respectively.