CAS RN 174568-92-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glyoxalase I inhibitor free base, MedChemExpress
MedChemExpress Glyoxalase I inhibitor (free base) is a potent Glyoxalase I (GLO1) inhibitor, candidate for anticancer agents.