CAS RN 173531-58-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Naphthoquine phosphate, MedChemExpress
MedChemExpress Naphthoquine phosphate is a potent and orally active antimalarial agent. Naphthoquine phosphate has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.