Learn More
Naphthoquine phosphate, MedChemExpress
Código de producto. 16491826
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
500 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
500 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16491826 500 mg 500 miligramos
16487236 100 mg 100 miligramos
16473297 50 mg 50 miligramos
16407288 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16491826 Proveedor Medchem Express N.º de proveedor HY17036
MedChemExpress Naphthoquine phosphate is a potent and orally active antimalarial agent. Naphthoquine phosphate has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.
Naphthoquine phosphate is a potent and orally active antimalarial agent. Naphthoquine phosphate has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.
Especificaciones
Nombre del producto químico o material Naphthoquine phosphate
Forma física Solid
CAS 173531-58-3
Color Amarillo claro
Cantidad 500 mg
Notas de grado de pureza Research
Fórmula molecular C24H34ClN3O9P2
Para utilizar con (aplicación) COVID-19-immunoregulation
Información de solubilidad H2O : 3.33 mg/mL (5.50 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.65 mM; Need ultrasonic)
SMILES ClC1=CC=C(C(NC2=C(CCCC3)C3=C(O)C(CNC(C)(C)C)=C2)=CC=N4)C4=C1.O=P(O)(O)O.O=P(O)(O)O
Peso molecular (g/mol) 605.94
Formula Weight (peso de la fórmula) 605.94
Porcentaje de pureza 98.0%