CAS RN 1702816-75-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Linperlisib, MedChemExpress
MedChemExpress Linperlisib (YY-20394) is a potent, orally bioavailable and selective inhibitor of PI3Kδ extracted from patent WO 2015055071 A1, compound 10; has an IC50 of 6.4 nM.