CAS RN 1699720-85-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(S)-ML753286, MedChemExpress
MedChemExpress (S)-ML753286 is a breast cancer resistance protein (BCRP) inhibitor with an IC50 of 0.6 μM on BCRP efflux transporter.