CAS RN 169815-40-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AQ-13 dihydrochloride, MedChemExpress
MedChemExpress AQ-13 dihydrochloride is an aminoquinoline antimalarial drug that is effective against drug-resistant strains of Plasmodium falciparum.