CAS RN 168161-71-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(Rac)-Sograzepide, MedChemExpress
MedChemExpress (Rac)-Sograzepide is an antagonist of cholecystokinin B (CCK-B) receptor, and has the potential of reducing the secretion of gastric acid.