CAS RN 1672665-49-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PT-2385, MedChemExpress
MedChemExpress PT-2385 is a selective HIF-2α inhibitor with a Ki of less than 50 nM.