CAS RN 1657014-42-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GDC-0575 dihydrochloride, MedChemExpress
MedChemExpress GDC-0575 dihydrochloride (ARRY-575 dihydrochloride) is an orally bioavailable CHK1 inhibitor, with an IC50 of 1.2 nM, and has antitumor activity.