CAS RN 1646299-50-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress Fmoc-Phe-Lys(Boc)-PAB-PNP
MedChemExpress Fmoc-Phe-Lys(Boc)-PAB-PNP is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).