CAS RN 1636881-61-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Mavoglurant racemate, MedChemExpress
MedChemExpress Mavoglurant racemate (AFQ-056 racemate) is the racemate of Mavoglurant. Mavoglurant is a novel, non-competitive mGlu5 receptor antagonist.