CAS RN 1633009-87-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BI 689648, MedChemExpress
MedChemExpress BI 689648 is a novel, highly selective aldosterone synthase inhibitor which can inhibit CYP11B1 and CYP11B2 with IC50s of 310 and 2.1 nM, respectively.