CAS RN 1628416-28-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CFTR corrector 2, MedChemExpress
MedChemExpress CFTR corrector 2 is a cystic fibrosis transmembrane conductance corrector (CFTR), extracted from patent US20140274933.