CAS RN 1628291-95-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HC-070, MedChemExpress
MedChemExpress HC-070 is an antagonist of TRPC4/TRPC5, with IC50s of 9.3 nM and 46 nM for hTRPC5 and hTRPC4 in cells, respectively.