CAS RN 1628210-26-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
KDM5-IN-1, MedChemExpress
MedChemExpress KDM5-IN-1 is a potent, selective and orally bioavailable KDM5 inhibitor with an IC50 of 15.1 nM.