CAS RN 1627607-88-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ (S)-PFI 2 hydrochloride
Negative control of (R)-PFI 2 hydrochloride (Cat. No. 4892)
2
Tocris Bioscience™ (S)-PFI 2 hydrochloride
Negative control of (R)-PFI 2 hydrochloride (Cat. No. 4892)