CAS RN 160312-62-9
IUPAC Nombre:
2-amino-N-[7,7-dimethyl-1-({[4-(2-methylphenyl)piperazin-1-yl]sulfonyl}methyl)bicyclo[2.2.1]heptan-2-yl]-4-methanesulfonylbutanamide hydrochloride
Sinónimos:
2-amino-N-(7,7-dimethyl-1-{[4-(2-methylphenyl)piperazin-1-ylsulfonyl]methyl}bicyclo[2.2.1]heptan-2-yl)-4-methanesulfonylbutanamide hydrochloride
Peso molecular (g/mol):
591.22
Fórmula molecular:
C26H43ClN4O5S2
InChi Key:
GIUFQWFJHXXXEQ-UHFFFAOYNA-N
SMILES:
Cl.CC1=CC=CC=C1N1CCN(CC1)S(=O)(=O)CC12CCC(CC1NC(=O)C(N)CCS(C)(=O)=O)C2(C)C
1
–
2
de
2
resultados
1
Tocris Bioscience™ L-368,899 hydrochloride
CAS: 160312-62-9 Fórmula molecular: C26H43ClN4O5S2
2
L-368,899 hydrochloride, MedChemExpress
MedChemExpress L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist, with IC50s of 8.9 nM and 26 nM for rat uterus and human uterus oxytocin receptor, respectively. L-368,899 hydrochloride used as a tocolytic agent.