CAS RN 15966-93-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SU5408, MedChemExpress
MedChemExpress SU5408 (VEGFR2 Kinase Inhibitor I) is a potent and cell-permeable inhibitor of VEGFR2 kinase with an IC50 of 70 nM.